missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Auxinole

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Auxinole is an auxin antagonist of TIR1/AFB receptors. Binds to TIR1 to block the formation of the TIR1-IAA-Aux/IAA complex and inhibit auxin-responsive gene expression. Auxinole competitively inhibits various auxin responses in planta. Auxinole also inhibits OsTIR1 that suppresses leaky degradation of degron-fused proteins. Can be used as a control compound with 5-Ph-IAA (Cat. No. 7392) in the AID2 degron system to switch off degradation.
Specifications
Specifications
| Chemical Name or Material | α-[2-(2,4-Dimethylphenyl)-2-oxoethyl]-1H-indole-3-acetic acid |
| CAS | 86445-22-9 |
| Quantity | 50 mg |
| Target | Molecular Glues |
| Molecular Formula | C20H19NO3 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction