missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AMG 853
GREENER_CHOICE

Product Code. 30000887
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30000887

Brand: Tocris Bioscience™ 5701/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent dual CRTH2 and prostaglandin D2 (PGD2) receptor antagonist

Potent dual CRTH2 and prostaglandin D2 (PGD2) receptor antagonist (IC50 values are 8 and 35 nM, respectively in plasma). Inhibits PGD2-induced down modulation of CRTH2 on CD16- granulocytes in human whole blood as well as PGD2-induced cAMP response in platelets. Inhibits PGD2-induced airway constriction in vivo. Also inhibits BRD4 (Kd = 170 nM). Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1169483-24-2
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Molecular Formula C28H27Cl2FN2O6S
Formula Weight 609.49
Purity >98%
Molecular Weight (g/mol) 609.49
Chemical Name or Material 5-Chloro-4-[2-[[(2-chloro-4-cyclopropylphenyl)sulfonyl]amino]-4-[[(1,1-dimethylethyl)amino]carbonyl]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.