missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMG-47a, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
AMG-47a is a potent and orally active lymphocyte-specific protein tyrosine kinase (Lck) inhibitor, with an IC50 of 0.2 nM. AMG-47a also inhibits VEGF2, p38α, Jak3 and MLR and IL-2 with IC50s of 1 nM, 3 nM, 72 nM, 30 nM and 21 nM, respectively. AMG-47a has anti-inflammatory activity.
Specifications
Specifications
| Chemical Name or Material | AMG-47a |
| Physical Form | Solid |
| CAS | 882663-88-9 |
| Color | Light Yellow |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C29H28F3N5O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 34 mg/mL (63.48 mM) |
| SMILES | O=C(NC1=CC=CC(C(F)(F)F)=C1)C2=CC=C(C)C(C3=CC4=CN=C(NCCN5CCOCC5)N=C4C=C3)=C2 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction