missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMG-337, MedChemExpress
Description
AMG-337 is a potent and highly selective small molecule ATP-competitive MET kinase inhibitor. AMG 337 inhibits MET kinase activity with an IC50 of < 5nM in enzymatic assays.
Specifications
Specifications
| Chemical Name or Material | AMG-337 |
| CAS | 1173699-31-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H22FN7O3 |
| Quantity | 200 mg |
| Solubility Information | DMSO : 100 mg/mL (215.77 mM; Need ultrasonic) |
| SMILES | O=C1C2=C(N=CC(OCCOC)=C2)C=CN1[C@@H](C3=NN=C4C(F)=CC(C5=CN(C)N=C5)=CN43)C |
| Molecular Weight (g/mol) | 463.46 |
| Formula Weight | 463.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?