missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMFR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16675-1-AP-150UL
This item is not returnable.
View return policy
Description
Autocrine motility factor is a tumor motility-stimulating protein secreted by tumor cells. The protein encoded by this gene is a glycosylated transmembrane protein and a receptor for autocrine motility factor. The receptor, which shows some sequence similarity to tumor protein p53, is localized to the leading and trailing edges of carcinoma cells.Specifications
| AMFR | |
| Polyclonal | |
| Unconjugated | |
| AMFR | |
| AMF receptor, isoform 2, AMFR, GP78, RING finger protein 45, RNF45 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23802, 267, 361367 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9R049, Q9UKV5 | |
| Amfr | |
| AMFR Fusion Protein Ag10178 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction