missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMFR/gp78 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | AMFR/gp78 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
AMFR/gp78 Polyclonal specifically detects AMFR/gp78 in Mouse samples. It is validated for Western Blot.Specifications
| AMFR/gp78 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS buffer, 2% sucrose | |
| 267 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| AMF receptor, AMF receptor, isoform 1, AMF receptor, isoform 2, autocrine motility factor receptor, Autocrine motility factor receptor, isoform 2, gp78, RING finger protein 45, RNF45EC 6.3.2.- | |
| The immunogen is a synthetic peptide directed towards the N terminal region of mouse AMFR/gp78 (NP_035917.2). Peptide sequence AYRALSQPEDGSGEPEPLTAPLQPEALAPARLTAGGPRARDVAQYLLSDS | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title