missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMFR/gp78 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38046-20ul
This item is not returnable.
View return policy
Description
AMFR/gp78 Polyclonal antibody specifically detects AMFR/gp78 in Human samples. It is validated for ELISA,Western Blot
Specifications
| AMFR/gp78 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| AMF receptor, AMF receptor, isoform 1, AMF receptor, isoform 2, autocrine motility factor receptor, Autocrine motility factor receptor, isoform 2, gp78, RING finger protein 45, RNF45EC 6.3.2.- | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 384-643 of human AMFR/gp78 (NP_001135.3).,, Sequence:, IADNNRVREEHQGENLDENLVPVAAAEGRPRLNQHNHFFHFDGSRIASWLPSFSVEVMHTTNILGITQASNSQLNAMAHQIQEMFPQVPYHLVLQDLQLTRSVEITTDNILEGRIQVPFPTQRSDSIRPALNSPVERPSSDQEEGETSAQTERVPLDLSPRLEETLDFGEVEVEPSEVEDFEARGSRFSKSADERQRMLVQRKDELLQQARKRFLNKSSEDDAASESFLPSEGASSDPVTLRRRMLAAAAERRLQKQQTS | |
| 20 μL | |
| Zinc Finger | |
| 267 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction