Learn More
Cytiva Amersham™ Streptavidin-HRP Conjugate
Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridization applications
Brand: Cytiva RPN1231-100UL
72.06 GBP valid until 2024-12-31
Use promo code "21954" to get your promotional price.
Code : GE
Additional Details : Weight : 0.01100kg
Description
- High specificity reagents
- Conjugation achieved using the periodate coupling method described by Nakane and co-workers
- Sold on the basis of titer in an ELISA and performance in a western blot
Specifications
Suitable for use in ECL™, western blotting ,ELISA, immunocytochemistry | |
2mL of Phosphate Buffered Saline (0.1M Sodium Phosphate, 0.1M Sodium Chloride, pH 7.4) containing 1%(w/v) Bovine Serum Albumin and an antimicrobial agent | |
100 mL |
Safety and Handling
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : reaction mass of: 5-chloro2-methyl-4-isothiazolin3-one ¢EC no. 247-500-7! and 2-methyl-2H-isothiazol