missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Streptavidin-Biotinylated HRP Complex

Product Code. 10470705 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10470705 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10470705 Supplier Cytiva Supplier No. RPN10512ML
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridization applications

  • High specificity reagents
  • Conjugation achieved using the periodate coupling method described by Nakane and co-workers
  • Sold on the basis of titer in an ELISA and performance in a western blot
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Suitable for use in ECL™, western blotting ,ELISA, immunocytochemistry
Includes 2mL of Phosphate Buffered Saline (0.1M Sodium Phosphate, 0.1M Sodium Chloride, pH 7.5) containing 1%(w/v) Bovine Serum Albumin and an antimicrobial agent
Quantity 2 mL
compliance-icons
Product Identifier
  • Streptavidin-Biotinylated Horseradish Peroxidase Complex, 2 ml
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: reaction mass of: 5-chloro2-methyl-4-isothiazolin3-one [EC no. 247-500-7] and 2-methyl-2H-isothiazol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.