Learn More
Cytiva Amersham™ Rediprime II DNA Labeling System
Long shelf-life and simple, fast protocol. Amersham™ Rediprime II DNA Labeling System consists of premixed, individually dispensed, random-prime reactions, freeze-dried for ambient stability.
Brand: Cytiva RPN1633
Code : GE
Additional Details : Weight : 0.10000kg
Description
p>Simply add substrate and [α-32P]dCTP. As little as 25ng substrate DNA can be used.- Ambient temperature-stable reactions with a color check for the labeling of DNA to specific activities of >109dpm/μg
- Long shelf life and simple, fast protocol; labeling reaction complete in 10 min.
- No need to remove unincorporated nucleotides before hybridization
- DNA can be labeled in solution or in the presence of low melting point agarose
Suitable for use with dCTP only.
Specifications
2°C to 25°C | |
Labeling reaction Buffered solution of dATP, dGTP, dTTP, Exonuclease-free Klenow Enzyme and random primers in a dried, stabilized form, control DNA 300ng of lambda Hind III DNA in a dried, stabilized form 1 tube | |
Rediprime II DNA Labeling System |
Labelling of DNA from a variety of sources to a high specific activity for use in Southern and Northern blot hybridizations | |
30 |
Safety and Handling
- Rediprime II, 30 reactions
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash hands thoroughly after handling.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : trometamol, 2-mercaptoethanol
- UNKNOWN ACUTE-20 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 20% of components with unknown hazards to the aquatic environment.