missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets

Product Code. 15299824 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10/Pk.
25/Pk.
5/Pk.
Length (Metric):
9 cm
10 cm
16 cm
20 cm
60 cm
Width (Metric):
8 cm
10 cm
14 cm
20 cm
30 cm
Pore Size:
0.2 μm
0.45 μm
Unit Size:
10 sheets
25 sheets
5 sheets
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Length (Metric) Width (Metric) Pore Size unitSize
15299824 25/Pk. 20 cm 20 cm 0.45 μm 25 sheets
15289814 5/Pk. 60 cm 30 cm 0.45 μm 5 sheets
15209834 10/Pk. 20 cm 20 cm 0.45 μm 10 sheets
15239844 25/Pk. 16 cm 14 cm 0.45 μm 25 sheets
15249854 10/Pk. 10 cm 10 cm 0.45 μm 10 sheets
15259854 25/Pk. 10 cm 10 cm 0.2 μm 25 sheets
15269854 10/Pk. 10 cm 10 cm 0.2 μm 10 sheets
15289864 25/Pk. 9 cm 8 cm 0.2 μm 25 sheets
15279864 25/Pk. 9 cm 8 cm 0.45 μm 25 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15299824 Supplier Cytiva Supplier No. 10600047

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Individual sheets.

 

  • Low background enables a dynamic range for complicated samples
  • Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability

 

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 20 cm
Width (Metric) 20 cm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Quantity 25/Pk.
Format Sheet
Pore Size 0.45 μm
compliance-icons
Product Identifier
  • Amersham Protran Premium
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.