missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Individual sheets.
- Low background enables a dynamic range for complicated samples
- Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
Specifications
Specifications
| Length (Metric) | 60 cm |
| Width (Metric) | 30 cm |
| For Use With (Application) | Western blots, dot-blot assays, and other protein or nucleic acid methods |
| Quantity | 5/Pk. |
| Format | Sheet |
| Pore Size | 0.45 μm |
Safety and Handling
Product Identifier
- Amersham Protran Premium
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?