Learn More
Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets
Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Marque: Cytiva 10600079
Code : GH
Informations supplémentaires : Weight : 0.00600kg
Description
Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Individual sheets.
- Low background enables a dynamic range for complicated samples
- Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
Spécification
10 cm | |
Western blots, dot-blot assays, and other protein or nucleic acid methods | |
Sheet |
10 cm | |
10/Pk. | |
0.45 μm |
Sécurité et traitement
- Amersham Protran Premium
Mention d'avertissement
- Warning
Catégorie de danger
- Flammable solid Category 2
Mention de danger
- H228-Flammable solid.
Conseils de prudence
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.