missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets

Product Code. 15239824 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10/Pk.
25/Pk.
5/Pk.
Length (Metric):
8 cm
10 cm
10.2 cm
16 cm
20 cm
60 cm
Width (Metric):
9 cm
10 cm
13.3 cm
14 cm
20 cm
30 cm
Pore Size:
0.1 μm
0.2 μm
0.45 μm
Unit Size:
10 sheets
25 sheets
5 sheets
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Length (Metric) Width (Metric) Pore Size Unit Size
15239824 25/Pk. 20 cm 20 cm 0.45 μm 25 sheets
15279814 5/Pk. 60 cm 30 cm 0.45 μm 5 sheets
15269814 5/Pk. 60 cm 30 cm 0.2 μm 5 sheets
15249824 10/Pk. 20 cm 20 cm 0.45 μm 10 sheets
15259824 25/Pk. 20 cm 20 cm 0.2 μm 25 sheets
15269824 10/Pk. 20 cm 20 cm 0.2 μm 10 sheets
15279824 25/Pk. 20 cm 20 cm 0.1 μm 25 sheets
15289824 10/Pk. 20 cm 20 cm 0.1 μm 10 sheets
15209844 25/Pk. 16 cm 14 cm 0.45 μm 25 sheets
15289844 10/Pk. 10 cm 10 cm 0.45 μm 10 sheets
15249864 25/Pk. 8 cm 9 cm 0.45 μm 25 sheets
15259864 25/Pk. 8 cm 9 cm 0.2 μm 25 sheets
15265928 10/Pk. 10.2 cm 13.3 cm 0.2 μm 10 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 15239824 Supplier Cytiva Supplier No. 10600041
Outlet Corner
Use promo code 28080
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Individual sheets.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 20 cm
Width (Metric) 20 cm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Quantity 25/Pk.
Format Sheet
Pore Size 0.45 μm
compliance-icons
Product Identifier
  • Amersham Protran
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.