missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls

Product Code. 15239794 Shop All Cytiva Products
Change view
Click to view available options
Width (Metric):
15 cm
20 cm
30 cm
Pore Size:
0.1 μm
0.2 μm
0.45 μm
Unit Size:
1 roll
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Pore Size unitSize
15239794 30 cm 0.1 μm 1 roll
15259794 30 cm 0.45 μm 1 roll
15249794 30 cm 0.2 μm 1 roll
15289794 20 cm 0.1 μm 1 roll
15299794 20 cm 0.2 μm 1 roll
15239804 15 cm 0.1 μm 1 roll
15259804 15 cm 0.45 μm 1 roll
15209804 20 cm 0.45 μm 1 roll
15249804 15 cm 0.2 μm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15239794 Supplier Cytiva Supplier No. 10600000
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

Packed in rolls.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 4 m
Width (Metric) 30 cm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Roll
Material 100% Pure Nitrocellulose
Pore Size 0.1 μm
compliance-icons
Product Identifier
  • Amersham Protran 0.1
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.