missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Megaprime™ DNA Labeling System

Product Code. 10575155 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
30 reactions
60 reactions
For Use With (Application):
dCTP Labeling
dNTP Labeling
Unit Size:
30 reactions
60 reactions
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity For Use With (Application) unitSize
10575155 60 reactions dNTP Labeling 60 reactions
10175804 60 reactions dCTP Labeling 60 reactions
10025174 30 reactions dNTP Labeling Each
10714367 30 reactions dCTP Labeling 30 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10575155 Supplier Cytiva Supplier No. RPN1605

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Primer solution; Labeling buffer; Reaction buffer; Enzyme; Standard DNA solution; Carrier DNA solution

For labeling DNA probes to specific activities >1 x 109dpm/μg in 10 minutes

Megaprime™ DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25ng substrate DNA.

  • Features cloned Klenow fragment and random nonamers in the random-prime labeling technique
  • No need to remove unincorporated nucleotides before hybridization
  • Protocols for labeling directly from low melting point agarose

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -15°C to -30°C
For Use With (Application) dNTP Labeling
Includes Primer solution 300μL, Labeling Buffer dATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2, Nucleotide solutions dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA 240μL, Reaction Buffer A 10 x concentra
No. of Reactions 60
Product Type Megaprime DNA Labeling System, dNTP (for use with any radioactively labeled nucleotide)
Quantity 60 reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.