Learn More
Cytiva Amersham™ Hybond™ -XL Membranes
Use for optimal signal-to-noise ratios when using radioactively labeled probes. Cytiva Life Sciences™ Amersham™ Hybond™ -XL Membranes are novel, charged nylon membranes that have a greater concentration and a more even distribution of positive charge than other charged nylon membranes.
Brand: Cytiva RPN2020S
Code : GE
Additional Details : Weight : 0.31900kg
Description
- Greater concentration and more even distribution of positive charge than other charged nylon membranes
- Binding capacity for nucleic acids up to 600μg/cm2
- Efficient binding of small fragments
- Effective probe removal
- Suitable for reprobing
- Discs are also available
Specifications
20 cm | |
Radioactive probes | |
10 Sheets | |
Nylon, Highly Positively Charged |
20 cm | |
Up to 600μg/cm2 | |
Sheet |