Learn More
Cytiva Amersham™ Hybond™ -N+ Membranes
Use for the highest sensitivity in DNA and RNA blotting. Cytiva Life Sciences™ Amersham™ Hybond™ -N+ Membrane is a positively charged nylon membranes for use with radioactive and nonradioactive chemiluminescence and chemifluorescence detection.
Brand: Cytiva RPN1782B
Code : GE
Additional Details : CAS Number : 156-57-0 Weight : 0.20200kg
Description
- Pore size of 0.45μm with retention of nucleic acids >50bp
- Binding capacity for nucleic acids 600μg/cm2
- Suitable for high-density arrays
- Excellent sensitivity in conventional Southern blotting
- Nucleic acid samples may be fixed by UV or baking
- Not suitable for alkali blotting and fixation
Specifications
Southern blots; UV fixation | |
82 mm | |
Gridded Disc | |
Not Applicable |
Up to 600μg/cm2 | |
50 Gridded Discs | |
Nylon, Positively Charged |