missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ Western Blotting Detection Reagents

Product Code. 10773577 Shop All Cytiva Products
Click to view available options
Quantity:
62.5 mL
125 mL
250 mL
375 mL
Sufficient For:
1000 cm2 Membrane
2000 cm2 Membrane
4000 cm2 Membrane
6000 cm2 Membrane
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10773577

Brand: Cytiva RPN2134

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Use for all routine Western blotting. Cytiva Life Sciences™ Amersham™ ECL™ Western Blotting Detection Reagents are the world's first commercially available chemiluminescent detection reagents and still one of the most widely used.

  • Detectable on Hyperfilm™ ECL or with CCD imager
  • Uses enhanced luminol-based detection which is suitable for all routine Western Blotting

Western Blotting

TRUSTED_SUSTAINABILITY

Especificaciones

Product Type ECL Western Blot Detection Reagent
For Use With (Application) Western Blot
Quantity 375 mL
Sensitivity ∼10 pg
Sufficient For 6000 cm2 Membrane
Kit Contents 375mL detection reagent, 1375mL detection reagent 2
Content And Storage 2°C to 8°C
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado