missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Fluorescent Nucleotides

Product Code. 10278184 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Cy3-CTP
Cy3-UTP
Cy3-dCTP
Cy3-dCTP, Cy5-dCTP
Cy3-dUTP
Cy3-dUTP, Cy5-dUTP
Cy3.5-dCTP
Cy5-CTP
Cy5-UTP
Cy5-dCTP
Cy5-dUTP
Cy5.5-dCTP
Quantity:
100 nm
100 nmol
25 nmol
250 nmol
5 x 25 nmol
Color:
Far-Red
Near IR
Orange
Orange, Far-Red
Scarlet
Unit Size:
1 set
Each
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Color unitSize
10278184 Cy3.5-dCTP 25 nmol Scarlet Each
10238804 Cy3-CTP 100 nm Orange Each
10554195 Cy5-CTP 100 nmol Far-Red Each
10743217 Cy3-UTP 100 nmol Orange Each
10713977 Cy3-dCTP 250 nmol Orange Each
10723977 Cy5-dUTP 250 nmol Far-Red Each
10126194 Cy5-UTP 100 nmol Far-Red Each
10095074 Cy3-dCTP, Cy5-dCTP 5 x 25 nmol Orange, Far-Red 1 set
10196094 Cy3-dUTP, Cy5-dUTP 5 x 25 nmol Orange, Far-Red 1 set
10279774 Cy5.5-dCTP 25 nmol Near IR Each
10712257 Cy3-dUTP 25 nmol Orange Each
10268614 Cy3-dCTP 25 nmol Orange Each
10627085 Cy5-dCTP 25 nmol Far-Red Each
10094684 Cy5-dUTP 25 nmol Far-Red Each
10441835 Cy5-dCTP 250 nmol Far-Red Each
10195844 Cy3-dUTP 250 nmol Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Product Code. 10278184 Supplier Cytiva Supplier No. PA53521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Label DNA and RNA probes. Cytiva Life Sciences™ Amersham™ CyDye™ Fluorescent Nucleotides are optimized for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.

For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.

  • Low nonspecific binding
  • High photostability
  • High water solubility
  • pH insensitive

TRUSTED_SUSTAINABILITY

Specifications

Description CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy dye labelled nucleotides exhibit the following properties:
Product Type Cy3.5-dCTP
Reagents Fluorescent Labeling Reagents
Quantity 25 nmol
Color Scarlet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.