missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ CyDye™ Fluorescent Nucleotides
Shop All Cytiva Products
Click to view available options
Product Type:
Cy3-CTP
Cy3-UTP
Cy3-dCTP
Cy3-dCTP, Cy5-dCTP
Cy3-dUTP
Cy3-dUTP, Cy5-dUTP
Cy3.5-dCTP
Cy5-CTP
Cy5-UTP
Cy5-dCTP
Cy5-dUTP
Cy5.5-dCTP
Quantity:
100 nm
100 nmol
25 nmol
250 nmol
5 x 25 nmol
Color:
Far-Red
Near IR
Orange
Orange, Far-Red
Scarlet
Unit Size:
1 set
Each
Description
For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.
- Low nonspecific binding
- High photostability
- High water solubility
- pH insensitive
Specifications
Specifications
| Description | CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy dye labelled nucleotides exhibit the following properties: |
| pH | 7 |
| Molecular Weight (g/mol) | 791.99 (Monofunction), 975.15 (Bisfunction) |
| Product Type | Cy5-dUTP |
| Reagents | Fluorescent Labeling Reagents |
| Wavelength | 670nm |
| Content And Storage | -15°C to -30°C |
| Quantity | 25 nmol |
| Color | Far-Red |
| For Use With (Application) | Genomic and Protein research, labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping and gene mapping |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction