missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMER2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558794
This item is not returnable.
View return policy
Description
Immunogen sequence: VDDTYLQEFW DMLSQTEEQG PEPQEGAAKV AAALETKVVP ETPKDTRCVE AAKDASSVKR RRLNRIPIEP HPKEEPKHPE KEQQE Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.
Negative regulator of the canonical Wnt signaling pathway involved in neuroectodermal patterning. Acts by specifically binding phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2), translocating to the cell membrane and interacting with key regulators of the canonical Wnt signaling pathway, such as components of the beta-catenin destruction complex.
Specifications
| AMER2 | |
| Polyclonal | |
| Unconjugated | |
| AMER2 | |
| 2600011E07Rik; AMER2; APC membrane recruitment 2; APC membrane recruitment protein 2; Fam123a; family with sequence similarity 123, member A; family with sequence similarity 123A; protein FAM123A; RGD1559605 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 219287 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N7J2 | |
| AMER2 | |
| Recombinant protein corresponding to Human AMER2. Recombinant protein control fragment (Product #RP-97993). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction