missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMER2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571433
This item is not returnable.
View return policy
Description
AMER2 Polyclonal Antibody for Western Blot
Negative regulator of the canonical Wnt signaling pathway involved in neuroectodermal patterning. Acts by specifically binding phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2), translocating to the cell membrane and interacting with key regulators of the canonical Wnt signaling pathway, such as components of the beta-catenin destruction complex.
Specifications
| AMER2 | |
| Polyclonal | |
| Unconjugated | |
| AMER2 | |
| 2600011E07Rik; AMER2; APC membrane recruitment 2; APC membrane recruitment protein 2; Fam123a; family with sequence similarity 123, member A; family with sequence similarity 123A; protein FAM123A; RGD1559605 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 219287 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N7J2 | |
| AMER2 | |
| synthetic peptide directed towards the middle region of human AMER2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction