missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMELY Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 85%; Dog: 85%; Goat: 85%; Horse: 85%; Human: 100%; Mouse: 77%; Pig: 92%; Rat: 85%; Sheep: 85%.
AMELY plays a role in the biomineralization of teeth. Seems to regulate the formation of crystallites during the secretory stage of tooth enamel development. Thought to play a major role in the structural organization and mineralization of developing enamel.
Specifications
Specifications
| Antigen | AMELY |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | AMELY |
| Gene Accession No. | Q99218 |
| Gene Alias | AMEL; Amelogenin; amelogenin (Y chromosome); Amelogenin 173A/173B; amelogenin 173B; amelogenin, Y isoform; amelogenin, Y-linked; AMELY; AMGL; AMGY; Class II amelogenin; Leucine-rich amelogenin peptide; LRAP; Y-linked amelogenin |
| Gene Symbols | AMELY |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?