missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AMELX Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5122244

 View more versions of this product

Product Code. 17723644

  • £155.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AMELX Polyclonal Antibody for Western Blot

This gene encodes a member of the amelogenin family of extracellular matrix proteins. Amelogenins are involved in biomineralization during tooth enamel development. Mutations in this gene cause X-linked amelogenesis imperfecta. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AMELX
Polyclonal
Unconjugated
AMELX
AI1E; AIH1; ALGN; Amel; amelogenesis imperfecta 1; amelogenin (amelogenesis imperfecta 1, X-linked); amelogenin (X chromosome, amelogenesis imperfecta 1); amelogenin X chromosome; amelogenin, X isoform; amelogenin, X-linked; Amelx; AMG; AMGL; AMGX; enamel matrix protein; leucine-rich amelogenin peptide; LRAP; M100888; Rgsc888
Rabbit
Antigen affinity chromatography, Protein A
RUO
265
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q99217
AMELX
Recombinant protein Amelogenin, X-Linked. The antigen corresponds to amino acid range 49-187 of the target protein.
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.