missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544172
This item is not returnable.
View return policy
Description
Peptide sequence: MGRMNSDCWY LYTLDFPESR VISQPDQTLE ILMSELDPAV MDQFYMKDGV Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
This gene encodes an important intermediate enzyme in polyamine biosynthesis. The polyamines spermine, spermidine, and putrescine are low-molecular-weight aliphatic amines essential for cellular proliferation and tumor promotion. Two alternatively spliced transcript variants that encode different proteins have been identified.
Specifications
| AMD1 | |
| Polyclonal | |
| Unconjugated | |
| AMD1 | |
| 1; adenosylmethionine decarboxylase 1; AdoMet decarboxylase; adoMetDC; AdoMetDC 1; adoMetDC1; AMD; Amd1; Amd-1; Amd1a; Amd1b; DKFZp313L1234; FLJ26964; S-adenosylmethionine decarboxylase; S-adenosylmethionine decarboxylase 1; S-adenosylmethionine decarboxylase 1 alpha chain; S-adenosylmethionine decarboxylase 1 beta chain; S-adenosylmethionine decarboxylase 1A; S-Adenosylmethionine decarboxylase 1B; S-adenosylmethionine decarboxylase alpha chain; S-adenosylmethionine decarboxylase beta chain; S-adenosylmethionine decarboxylase proenzyme; S-adenosylmethionine decarboxylase proenzyme 1; S-adenosylmethionine decarboxylase proenzym-like protein; SAMDC; SAMDC 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 262 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P17707 | |
| AMD1 | |
| Synthetic peptide directed towards the N-terminal of human AMD1 (aa 149-198). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction