missing translation for 'onlineSavingsMsg'
Learn More

AMD 3465 hexahydrobromide, Tocris Bioscience™

Product Code. 13434294
Encompass
Click to view available options
10mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective CXCR4 antagonist

Potent, selective CXCR4 antagonist; exhibits 8-fold higher affinity than AMD 3100 (Cat.No. 3299). Inhibits SDF-1α-ligand binding (Ki = 41.7 nM). Potently inhibits HIV cell entry in vitro; causes leukocytosis and mobilizes haematopoietic stem cells in vivo.

Specifications

CAS 185991-07-5
Molecular Formula C24H44Br6N6
Quantity 10mg
Synonym amd 3465 hexahydrobromide, pyridin-2-ylmethyl 4-1,4,8,11-tetraazacyclotetradecan-1-ylmethyl phenyl methyl amine hexahydrobromide, n-4-1,4,8,11-tetraazacyclotetradec-1-ylmethyl phenyl methyl-2-pyridinemethanamine hexahydrobromide, amd3465 hexahydrobromide, amd3465 6 hbr, n-pyridin-2-ylmethyl-1-4-1,4,8,11-tetrazacyclotetradec-1-ylmethyl phenyl methanamine hexahydrobromide
InChI Key ARHBIBDGWDRBJH-UHFFFAOYSA-N
SMILES C1CNCCNCCCN(CCNC1)CC2=CC=C(C=C2)CNCC3=CC=CC=N3.Br.Br.Br.Br.Br.Br
IUPAC Name N-(pyridin-2-ylmethyl)-1-[4-(1,4,8,11-tetrazacyclotetradec-1-ylmethyl)phenyl]methanamine;hexahydrobromide
Molecular Weight (g/mol) 896.082
PubChem CID 9897616
Formula Weight 896.07
Percent Purity >97%
Chemical Name or Material AMD 3465 hexahydrobromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'