missing translation for 'onlineSavingsMsg'
Learn More

Amcasertib, MedChemExpress

Product Code. 16483437
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483437 50 mg 50mg
16441709 1 mg 1mg
16417418 10 mg 10mg
16409897 5 mg 5mg
16407396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483437 Supplier Medchem Express Supplier No. HY17602
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amcasertib (BBI503) is an orally active and small-molecule multi-kinase inhibitor. Amcasertib exhibits inhibitory activity against the NANOG and CD133 expression and cell viability in PC-9/GR cells. As an orally available cancer cell stemness kinase inhibitor with potential antineoplastic activity, it is currently being studied in phase I clinical trials in a number of cancers.

Amcasertib (BBI503) is an orally active and small-molecule multi-kinase inhibitor. Amcasertib exhibits inhibitory activity against the NANOG and CD133 expression and cell viability in PC-9/GR cells. As an orally available cancer cell stemness kinase inhibitor with potential antineoplastic activity, it is currently being studied in phase I clinical trials in a number of cancers.

Specifications

Chemical Name or Material Amcasertib
CAS 1129403-56-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H33N5O2S
Quantity 50 mg
Synonym BBI503
Solubility Information DMSO : ≥ 30 mg/mL (55.59 mM)
SMILES O=C(C1=C(C)NC(/C=C2C(NC3=C/2C=C(C4=CSC(C5=CC=CC=C5)=N4)C=C3)=O)=C1C)NCCN(CC)CC
Molecular Weight (g/mol) 539.69
Formula Weight 539.69
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.