missing translation for 'onlineSavingsMsg'
Learn More

Ambrisentan, TRC

Product Code. 30315044
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30315044

Brand: Toronto Research Chemicals TRCA57586050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ambrisentan
CAS 177036-94-1
Molecular Formula C22 H22 N2 O4
Quantity 50 mg
Synonym (αS)-α-[(4,6-Dimethyl-2-pyrimidinyl)oxy]-β-methoxy-β-phenyl-benzenepropanoic Acid, Benzenepropanoic acid, α-[(4,6-dimethyl-2-pyrimidinyl)oxy]-β-methoxy-β-phenyl-, (S)-, (αS)-α-[(4,6-Dimethyl-2-pyrimidinyl)oxy]-β-methoxy-β-phenylbenzenepropanoic acid, Ambrisentan, BSF 208075, LU 208075, Letairis, Volibris
InChI Formula InChI=1S/C22H22N2O4/c1-15-14-16(2)24-21(23-15)28-19(20(25)26)22(27-3,17-10-6-4-7-11-17)18-12-8-5-9-13-18/h4-14,19H,1-3H3,(H,25,26)/t19-/m1/s1
SMILES COC([C@H](Oc1nc(C)cc(C)n1)C(=O)O)(c2ccccc2)c3ccccc3
IUPAC Name (2S)-2-(4,6-dimethylpyrimidin-2-yl)oxy-3-methoxy-3,3-diphenylpropanoic acid
Molecular Weight (g/mol) 378.42
Formula Weight 378.158
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.