missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambrisentan, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Ambrisentan
Specifications
Specifications
| Chemical Name or Material | Ambrisentan |
| CAS | 177036-94-1 |
| Analyte or Component Names | Ambrisentan |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H22 N2 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Ambrisentan, BSF 208075, LU 208075, Letairis, Volibris, (alphaS)-alpha-[(4,6-Dimethyl-2-pyrimidinyl)oxy]-beta-methoxy-beta-phenyl-benzenepropanoic Acid, Benzenepropanoic acid, alpha-[(4,6-dimethyl-2-pyrimidinyl)oxy]-beta-methoxy-beta-phenyl-, (S)-, (alphaS)-alpha-[(4,6-Dimethyl-2-pyrimidinyl)oxy]-beta-methoxy-beta-phenylbenzenepropanoic acid, 3109.F, GSK 1325760, 4051.F |
| InChI Formula | InChI=1S/C22H22N2O4/c1-15-14-16(2)24-21(23-15)28-19(20(25)26)22(27-3,17-10-6-4-7-11-17)18-12-8-5-9-13-18/h4-14,19H,1-3H3,(H,25,26)/t19-/m1/s1 |
| SMILES | COC([C@H](Oc1nc(C)cc(C)n1)C(=O)O)(c2ccccc2)c3ccccc3 |
| IUPAC Name | (2S)-2-(4,6-dimethylpyrimidin-2-yl)oxy-3-methoxy-3,3-diphenylpropanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction