Learn More
Invitrogen™ Water-Saturated Phenol, pH 6.6
Description
Ambion Water Saturated Phenol is supplied premixed and at pH 6.6 (pH 7.9 ±0.2, with included buffer). Water Saturated Phenol is used in RNA and DNA extraction procedures. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality tested, saturated phenols are ready-to-use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Chemical Name or Material | Phenols |
| Content And Storage | Store at 4°C or –20°C. |
| Packaging Type | Bottle |
| pH | 6.6 |
| Product Line | Ambion |
| Purity | Molecular Biology Grade |
| Quantity | 100 mL |
| Shipping Condition | Room Temperature |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.