missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Buffer-Saturated Phenol

Product Code. p-4926944
Change view
Click to view available options
Quantity:
100 mL
400mL
Unit Size:
100mL
400mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11598886 100 mL 100mL
11508896 400mL 400mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11598886 Supplier Invitrogen™ Supplier No. 15513039
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

UltraPure™ Buffer-Saturated Phenol is used in the purification of nucleic acids. The reagent, which consists of UltraPure™ Phenol that has been saturated with Tris-HCl buffer, is already buffer equilibrated to pH >7.4.

  • UltraPure™ Buffer-Saturated Phenol is used in the purification of nucleic acids
  • The reagent, which consists of UltraPure™ Phenol that has been saturated with Tris-HCl buffer, is already buffer equilibrated to pH >7.4
  • When mixtures are extracted with UltraPure™ Buffer- Saturated Phenol, proteins are denatured and collect in the organic phase or at the interphase, while most nucleic acids remain in the aqueous phase
  • UltraPure™ Buffer-Saturated Phenol contains no preservatives
  • It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles
  • The appearance of the solution is evaluated at room temperature
  • No RNase or DNase activity detected
  • Store in refrigerator at 2°C to 8°C

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenols
Content And Storage Store in refrigerator (2–8°C).
Packaging Type Bottle
pH 7.4
Product Line UltraPure
Purity Molecular Biology Grade
Quantity 100 mL
Shipping Condition Room Temperature
Form Liquid
Do you have any information on DNA and RNA purification using phenol chloroform and alcohol precipitation?

Phenol extraction of proteins:

Phenol extraction is frequently used to remove proteins from nucleic acid solutions. A common protocol is to add an equal volume of buffer-saturated phenol or phenol:chloroform:isoamyl alcohol (25:24:1, v/v/v) to an aqueous nucleic acid solution, vortex, and centrifuge at 14,000 x g for 1 min to separate the phases.

Studies at Thermo Fisher Scientific have shown that the concentration of NaCl in the aqueous solution should not exceed 0.5 M for good recovery of DNA. Residual phenol can be removed from the aqueous phase by extraction with an equal volume of chloroform or ether. After extraction, DNA is usually precipitated with ammonium acetate and ethanol as described in another protocol on this server. Ref. Karger, B. D. (1989) FOCUS 11, 14.

A good source of general information on the properties of phenol can be found in Wallace, Donald M. “Large and Small-Scale Phenol Extractions”. Methods in Enz. Volume 152 guide to Molecular Cloning Techniques. 1987. Academic Press, Inc. Berger and Kimmel, eds. Chap.4, pg 33-41.

(a) At pH 5 to 6 DNA is selectively retained in the organic phase and interphase, leaving RNA in the aqueous phase. Therefore a pH greater than 7 is needed if DNA is to be extracted.

(b) At pH values below 7.6, poly A+ RNA is lost to the organic phase if chloroform is not present.

(c) Optimal RNA yields in phenol extraction are obtained if the salt concentration is less than 0.15 M NaCl. Salt concentration in the sample is not a factor for larger DNA molecules.

To store RNA after extraction use DEPC-treated water.

What is the recommended protocol for phenol-extraction removal of proteins from nucleic acid containing solutions?

Below is a commonly used protocol:

(1) Add an equal volume of buffer-saturated phenol or phenol:chloroform:isoamyl alcohol (25:24:1, v/v/v) to an aqueous nucleic acid solution. Note: for RNA solutions, acid-phenol is recommended.

(2) Vortex, and centrifuge at 14,000 x g for 1 min to separate the phases.

(3) Residual phenol can be removed from the aqueous phase by extraction with an equal volume of chloroform or ether.

(4) After extraction, DNA/RNA is usually precipitated with ammonium acetate and ethanol.

What is the optimal pH of the phenol:chloroform mixture for isolation of DNA?

Partitioning of the nucleic acids in phenol is pH dependent. At pH 7.0 or higher, both DNA and RNA partition into the aqueous phase. At an acidic pH (below 7.0) DNA is denatured and will move into the organic phase, but the RNA remains in the aqueous phase. The mixture should be adjusted to at least pH 7.4 for work with DNA.

Recently I came across a DNA purification technique, which uses urea during phenol extraction. What is the purpose of using urea?

Using urea during phenol extraction denatures the protein associated with the DNA and the proteins that bind the genomic DNA to the cell wall.

compliance-icons compliance-icons compliance-icons
Product Identifier
  • Buffer-Saturated Phenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: phenol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.