missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ULTRAhyb™ Ultrasensitive Hybridization Buffer with Manual

Product Code. 10076334
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10076334 125 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10076334 Supplier Invitrogen™ Supplier No. AM8670M

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Proprietary blend of hybridization accelerators and blocking agents to enhance the levels of hybridization

  • Signals that once took days to visualize are now apparent in hours
  • It is supplied in four bottles containing 125mL each
  • Increase the sensitivity of any blot hybridization experiment
  • Increase random-primed DNA probe sensitivity up to 100x; RNA probe sensitivity 20x
  • Decrease hybridization time to 2 hours
  • Using standard hybridization buffers, only 1 to ;5% of target molecules on a blot hybridizes to probe, making blots a relatively insensitive method for nucleic acid analysis (Vernier et al. (1996) Anal Biochem 235: 11-19): With ULTRAhyb™ Ultrasensitive Hybridization Buffer, the hybridization reaction approaches completion, so that as few as 10,000 molecules can be detected
  • Compatible with RNA and DNA probes labeled isotopically and nonisotopically in northerns, Southerns, and dot/slot blots
  • Contains 50% formamide and is compatible with positively charged membranes
  • Requires 10-100x less nonisotopic DNA probe than standard hybridization buffers, making it possible to perform many more hybridizations with a single labeling reaction

Applications:

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

TRUSTED_SUSTAINABILITY

Specifications

Buffer Hybridization
Quantity 125 mL
Storage Requirements 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.