missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ULTRAhyb™-Oligo

Product Code. 10457304
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10457304 125 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10457304 Supplier Invitrogen™ Supplier No. AM8663

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Specifically formulated for use with oligonucleotide probes in northern analysis

  • Contains a proprietary blend of hybridization accelerators and blocking agents to enhance the levels of hybridization
  • Supplied in one bottle containing 125mL
  • Use less probe per blot
  • Decrease hybridization time to 2 hours
  • Using standard hybridization buffers, only 1 to 5% of target molecules on a blot hybridizes to probe, making blots a relatively insensitive method for nucleic acid analysis (Vernier et al. (1996) Anal Biochem 235: 11-19)
  • With ULTRAhyb™ Ultrasensitive Hybridization Buffer, the hybridization reaction approaches completion, so that as few as 10,000 molecules can be detected
  • It works well with oligonucleotides having Tms from 78° to 90°C and 47 to 62% GC contents; However, the optimal hybridization temperature for oligonucleotide probes with different GC content and Tm values should be determined empirically

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Buffer Hybridization Buffer
Quantity 125 mL
Shipping Condition Room Temperature
Product Type Oligo Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.