Learn More
Invitrogen™ Ambion™ TURBO DNA-free Kit with Manual
Description
TURBO DNA-free™ Kit contains reagents for efficient, complete digestion of DNA along with removal of enzyme and divalent cations post-digestion
Includes
- Catalytically superior enzyme compared to wild type DNase I
- Removes trace quantities of DNA that can interfere with RT-PCR
- Reagent included to completely remove DNase without phenol treatment or heating
- Method for genomic DNA removal prior to RT-PCR
- Efficient DNase and divalent cation removal without organic extraction or precipitation
PCR & Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription
Specifications
Specifications
| Content And Storage | Store TURBO DNase, 10X TURBO DNase Buffer and DNase Inactivation Reagent at -20°C. Store Nuclease–free Water at room temperature. |
| Quantity | 50 Reactions |
| Product Type | Kit with manual |