missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Green Cells-to-CT™ Control Kit
GREENER_CHOICE

This kit is designed for use with the Power SYBRGreen and Fast SYBRGreen Cells-to-CT™ Kits with sufficient reagents are provided for 100 reactions.

Brand:  Invitrogen™ 4402959

144.67 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.


Product Code. 10637705

  • £217.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price


Includes: 100μL Xeno RNA Control, 105 copies/μL; 250μL 20X SYBR Xeno Primers; 250μL 20X SYBR ACTB Primers; Amounts: Xeno RNA Control: approximately 100 lysis reactions, SYBR Xeno and SYBR ACTB Primers: up to 250 PCRs (20μL) each
This item is not returnable. View return policy

Description

Description

This kit is designed for use with the Power SYBRGreen (Cat. No. 4402953, 4402954, and 4402955) and Fast SYBRGreen Cells-to-CT™ Kits (Cat. No. 4402956 and 4402957). Sufficient reagents are provided for 100 reactions. It includes Xeno™ RNA Control, a synthetic RNA transcript with a unique sequence that lacks homology to current annotated biological sequences, and a PCR primer set for the Xeno RNA Control target. It also includes a PCR primer set for the highly expressed endogenous control gene β-actin (ACTB). Together these reagents provide positive controls for reverse transcription and SYBRGreen-based real-time PCR, and can indicate the presence of any reverse transcription or PCR inhibitors. Real-time RT-PCR with SYBRACTB Primers can also provide an endogenous control for sample normalization, as well as to confirm sufficient cell input in experiments with samples consisting of more than 100 cells per lysis.

  • Polymerase: AmpliTaq Gold™ DNA Polymerase
  • Purity/Quality Grade: UP (Ultra Pure)

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Components:
• 100 μL Xeno™ RNA Control, 105 copies/μL
• 250 μL 20X SYBR Xeno Primers
• 250 μL 20X SYBR ACTB Primers

Amounts:
• Xeno RNA Control: approximately 100 lysis reactions
• SYBR Xeno and SYBR ACTB Primers: up to 250 x 20 μL PCRs

Storage Conditions: Store at −20°C. Do not store in a frost-free freezer.

Tube
Fewer resources used and less waste, Less hazardous
Cells-to-CT™, SYBR™
Dry Ice
100 Reactions
XenoRNA™ control (synthetic RNA transcript)
ACTB
Unconjugated
100 Reactions
XenoRNA™ control, β-actin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.