Learn More
Ambion™ SuperTaq™ Polymerase (Cloned) 5 U/μl
High-purity polymerase isolated from an E. coli strain that over-expresses this gene
Brand: ABI Applied Biosystems AM2052
Code : 50
Additional Details : Weight : 0.01000kg
Includes: dNTPs, 10X Reaction Buffer with MgCl2, 10X Reaction Buffer without MgCl2, and MgCl2are stored at -20°C.
Description
Exceptional heat stability allows DNA polymerization in the 5« to 3« direction during repeated heat denaturation and reannealing steps in the presence of dNTPs, a DNA template, and complementary primers.
- Supplied in tube containing 250U at concentration of 5U/μL for use in PCR and RT-PCR reactions
- Unit Definition:One unit incorporates 10nmol of deoxynucleotides into acid insoluble material in 30 minutes at 74°C
SuperTaq™ is a trademark of, and is manufactured by Enzyme Technologies Ltd. and sold under licensing arrangements with F. Hoffman-La Roche, Ltd., Roche Molecular Systems, Inc. and the Perkin-Elmer Corporation. Life Technologies Corporation is a distributor of Enzyme Technologies, Ltd., except in the following countries: Austria, Benelux, Denmark, Greece, Sweden, Switzerland, Taiwan, and the United Kingdom.
PCR, PCR and Real-Time PCR, Routine PCR
Order Info
Shipping Condition: Dry ice
Specifications
dNTPs, 10X Reaction Buffer with MgCl2, 10X Reaction Buffer with out MgCl2, and MgCl2. Store at –20°C. | |
SuperTaq Polymerase | |
250 units | |
Mixed |
Low | |
None | |
1 X | |
2.5 kb or less |
For Research Use Only. Not for use in diagnostic procedures.