Learn More
Invitrogen™ Ambion™ SP6 RNA Polymerase-Plus (20U/μL)
Description
Includes
One tube containing 1000 units at a concentration of 20U/μL
- This enzyme has a high specificity for its promoter, transcribing large amounts of RNA with no cross talk between promoters
- It is supplied in one tube containing 1000U at a concentration of 20U/μL
- The enzyme incorporates radiolabeled nucleotides along with many modified nucleotides
- Inclusion of RNase inhibitor provides broad-spectrum protection against enzymatic degradation of the synthesized RNA, making the enzyme ideal for the synthesis of RNA for blot and in situ hybridization probes, RPA analysis, in vitro translation, and antisense RNA
Unit Definition: One unit catalyzes the incorporation of 1nmole of NTP into acid-insoluble material in 60 minutes using a linearized template approximately 300 nt in length. Using a short linear template in the unit assay is essential because it allows the measurement of the rate limiting initiation step. Units determined from long (3000 nt) supercoiled templates appear to be 3- to 5-fold higher, but they measure the elongation rate that is much faster than the initiation rate. Thus, one unit defined with a linearized 300 nt template actually contains more polymerase activity than a unit defined using a 3000 nt supercoiled template.
DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, in vitro Transcription, Nucleic Acid Labeling & Oligo Synthesis, RNA Labeling
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Concentration | 20 U/μL |
| Content And Storage | One tube containing 1000 units at a concentration of 20U/μL. Store at -20deg.C |
| Quantity | 1000 units |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.