missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ RNase III

Product Code. 10301375
Change view
Click to view available options
Quantity:
250 units
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10301375 250 units Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10301375 Supplier Invitrogen™ Supplier No. AM2290

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Double-stranded RNA (dsRNA) specific endoribonuclease

Escherichia coli Ribonuclease III (RNase III; EC 3.1.24) is specific endoribonuclease. In E. coli, RNase III cleaves ribosomal RNA (rRNA) precursors during maturation of rRNA. Enzyme cleaves dsRNA into 12–15 bp dsRNA fragments with 2 to 3 nucleotide 3' overhangs, and 5' phosphate and 3' hydroxyl termini. Termini and overhangs of RNase III cleavage products are thus same as those produced by Dicer in eukaryotic RNAi pathway. Transfection of RNase III cleavage products can be used to induce RNAi in mammalian cells (patent pending).

  • Unit Definition: One unit is defined as amount of enzyme catalyzing the cleavage of 1μg of 500 bp dsRNA substrate to approximately 12–30 bp fragments in 60 min. at 37°C

DNA & RNA Purification & Analysis, In Vitro Transcribed siRNA & Dicer siRNA, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting, RNAi, RNAi, Epigenetics & Noncoding RNA Research

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage RNase III and 10X Reaction Buffer should be stored at –20°C.
Shipping Condition Dry Ice
Enzyme RNase
Compatible Buffer 10X Reaction Buffer
Quantity 250 units
Product Type RNase III
Product Line Ambion

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.