missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseZap™ RNase Decontamination Solution

Product Code. 10586925
Click to view available options
Quantity:
250 mL
4 L
6 x 250 mL
Product Type:
RNase
RNase (with manual)
Unit Size:
1500mL
250mL
Each
This item is not returnable. View return policy

Product Code. 10586925

Brand: Invitrogen™ AM9780M

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

RNaseZap™ RNase Decontamination Solution is a surface decontamination solution that destroys RNases on contact. You simply spray RNaseZap™ Solution onto the surface to be decontaminated and rinse it off with RNase-free water.

RNaseZap™ RNase Decontamination Solution is a surface decontamination solution that destroys RNases on contact. You simply spray RNaseZap™ Solution onto the surface to be decontaminated and rinse it off with RNase-free water.

Features of RNaseZap™ RNase Decontamination Solution:
• Completely removes RNase contamination from glass and plastic surfaces
• Excels at removing high levels of RNase contamination whereas similar products fail
• Proven effective at removing high concentrations of dried-on RNase A
• Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free

Using RNaseZap™ Solution
Working with RNA requires that special measures be taken to ensure an RNase-free environment. Even trace quantities of RNase can lead to lower yields from in vitro transcription reactions, degradation during RNA purification protocols, and variable results in RPAs and Northerns. RNaseZap™ contains three ingredients active against RNase and has proven to be extremely effective at removing RNase contamination from glassware, plastic surfaces, countertops, and pipettors. It has also been shown to be effective at eliminating RNase contamination from microfuge tubes without inhibiting subsequent enzymatic reactions. Supplied in one 250 mL bottle.

RNase contamination in microfuge tubes
While concern about RNase contamination of frequently handled labware is common and prudent, researchers often do not worry about RNase contamination in commercially prepared products such as microfuge tubes. However, in a survey of six standard 1.5 mL microfuge tubes in which three were pre-rinsed with RNaseZap™ and water and three were left untreated, all of the untreated tubes had at least marginal levels of RNase contamination, whereas none of the tubes pre-rinsed with RNaseZap™ exhibited contamination. Additionally, in testing commercially available “RNase-free” microfuge tubes, approximately 10% had some RNase contamination (data not shown). This may well account for the erratic results that researchers periodically experience when performing RNA-based assays.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type RNase (with manual)
Reagents RNase Control Reagent
Quantity 250 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.