Learn More
Invitrogen™ Ambion™ RNase A (RPA grade) 1mg/mL
Endonuclease-free, exonuclease-free, protease-free
Brand: Invitrogen™ AM2272
Code : 50
Additional Details : Weight : 0.10500kg
Description
RPA Grade RNase A is intended for those who desire to use RNase A alone in ribonuclease protection assays instead of usual mixture of RNases A and T1. This product is equivalent to molecular biology grade RNase A from other suppliers.
- Endonuclease that specifically cleaves 3' of U and C residues
- Supplied in one tube containing 1mg (1mg/mL)
- Free of DNase and nicking activities
- Rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity and functionality is determined in a standard RPA III™ (Mfr. No. AM1414) reaction
DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Midiprep, Miniprep, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification
Order Info
Shipping Conditions: Dry ice
Specifications
-20°C | |
1 mg |
For Research Use Only. Not for use in diagnostic procedures.