missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNAqueous™-96 Total RNA Isolation Kit

Product Code. 10137304
Change view
Click to view available options
Quantity:
192 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10137304 192 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10137304 Supplier Invitrogen™ Supplier No. AM1920

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The RNAqueous™-96 Kit is used for high-throughput, phenol-free total RNA isolation from 100–2,000,000 cells or 0.1–1.5 mg of tissue using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.

The RNAqueous™-96 Kit is used for high-throughput, phenol-free total RNA isolation from 100–2,000,000 cells or 0.1–1.5 mg of tissue using a guanidinium-based lysis/denaturant and glass fiber filter separation technology. The kit contains sufficient reagents for 192 (2 x 96) RNA purifications. The RNAqueous™-96 Kit is ideal for the isolation of total RNA from multiple samples and very small samples.

Using the RNAqueous™-96 Kit
The procedure uses an RNA-binding glass-fiber filter and either vacuum filtration or centrifugation methods to provide high yields of intact RNA. An optional on-the-filter DNase treatment can be performed to ensure removal of genomic DNA for RT-PCR applications. The 96-well plates are compatible with most common 96-well plate vacuum manifolds and 96-well plate centrifuge rotors. The RNAqueous™-96 Kit provides consistently high yields of RNA from various amounts of starting material. The high quality of RNA isolated is illustrated (see figure).

Accessory product
The RNAqueous™-96 Automated Kit (Cat. No. AM1812) is the same product in a format suitable for automation.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 50 mL Nuclease-free Water; store at room temperature
• 50 mL Elution Solution; room temperature
• 2 Filter Plates; room temperature
• 2 Collection Plates; room temperature
• 2 Adhesive Plate Sealers; room temperature
• 2 Plate Lids; room temperature
• 50 mL Lysis/Binding Solution; 4°C
• 175 mL Wash Solution (concentrated); 4°C
• 53 mL Rebinding Mix; 4°C
• 12 mL DNase I Buffer; -20°C
• 1.25 mL DNase I; -20°C
Isolation Technology Silica Plate
Elution Volume ≥50 μL
Sample Type Cells, Tissue
Final Product Type Total RNA
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) MultiPROBE™ II HT
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 192 Preps
Shipping Condition Box 1: Dry Ice
Box 2: Room Temperature
Starting Material Amount Cells: ≤106
Tissue: ≤1.5 mg
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.