missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNAsecure™ Resuspension Solution (1 ml Tube)

Product Code. 10782067
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10782067 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10782067 Supplier Invitrogen™ Supplier No. AM7010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Non-enzymatic reagent inactivate RNases in solution

Invitrogen RNAsecure™ Resuspension Solution (patents pending) is a unique non-enzymatic reagent that will irreversibly inactivate RNases in solution. It is supplied at working concentration for direct resuspension of RNA pellets. Ten tubes containing 1mL each are provided. RNAsecure™ Resuspension Solution is specifically meant for resuspending RNA pellets that may contain trace amounts of ribonuclease (e.g., after total RNA isolation). Note: it cannot reverse RNA degradation that has already occurred, but can prevent further RNA degradation. To use, the RNA pellet is resuspended in the RNAsecure Resuspension Solution and heated to 60°C for 10 minutes It can be reheated to eliminate any RNase contamination introduced at a later time.

  • No post-treatment autoclaving
  • Safe, easy-to-use reagent inactivates RNases in solution
  • Inactivation process can be repeated to protect against newly introduced contaminants
  • Compatible with downstream procedures, such as RT-PCR and in vitro transcription

DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, PCR and Real-Time PCR, RNA Extraction, Real Time PCR (qPCR), Total RNA Isolation, Total RNA from Animal Cells and Tissues, mRNA Isolation

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store pure solution at –20°C.
Packaging Type Tubes
Product Line Ambion, RNAsecure
Product Type RNase Control Reagent
Purity Molecular Biology Grade
Quantity 1 mL
Shipping Condition Dry Ice
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.