Learn More
Invitrogen™ RiboPure™ RNA Purification Kit, bacteria
High quality RNA isolation from bacteria
Brand: Invitrogen™ AM1925
Code : 50
Additional Details : CAS Number : 108-95-2 Weight : 0.68000kg Transport : UN number : 2821 Chem class : 6.1 Pack group : II
Description
The Invitrogen RiboPure-Bacteria Kit provides a rapid, robust, column-based method for isolating high quality RNA from bacteria. Total RNA can be isolated from the toughest Gram positive bacteria samples thanks to the kit's comprehensive protocol that uses zirconia beads to thoroughly disrupt bacterial cell walls. The RiboPure-Bacteria Kit protocol also eliminates the need for harsh treatments or enzymatic digestion which may alter gene expression profile.
A Comprehensive and Convenient Column Purification Procedure The RiboPure-Bacteria Kit purifies total RNA from a wide range of bacterial species and has been extensively tested with E. coli, Pseudomonas aeruginosa, Staphylococcus aureus, Bacillus subtilis, Campylobacter fetus, and Rhodobacter sphaeroides.
A Streamlined and Effective Method for Bacterial RNA Extraction The kit protocol begins with thorough cell wall disruption using of zirconia beads, followed by organic extraction of the lysate, and RNA purification on a convenient silica column. In approximately one hour, RNA is of superb quality can be isolated from fresh, snap-frozen, or RNAlater solution-treated bacterial cells. To help ensure complete removal of genomic DNA, an optional DNA removal step requiring approximately 30 minutes of additional time, can be incorporated. When the RiboPure-Bacteria kit is used upstream from theMICROBExpress Bacterial mRNA Enrichment Kit, the sensitivity of RNA-Seq experiments or array analyses can be dramatically increased.
Purified RNA is Suitable for Use with a Variety of Downstream Applications and Products: The RNA purified using the Invitrogen RiboPure-Bacteria Kit is suitable for downstream applications including RNA sequencing, microarray analysis, real-time PCR (qPCR), reverse transcription qPCR (RT-qPCR), northern blotting, and cDNA library construction.
- Obtain purified bacterial RNA suitable for nearly any downstream application
- Prepare pure RNA samples from the most difficult-to-lyse Gram positive bacteria
- Eliminate enzymatic pretreatments that might otherwise alter gene expression
- Remove contaminating gDNA with included DNA-free™ reagents
Accessory Products: The Kit requires the use of the Vortex Adapter (Cat. No. AM10024), which turns a standard laboratory vortex into a bead beater. This adapter fits the Vortex Genie™ 2 and holds 12 tubes.
Not high-throughput compatible (Manual)
Applications:
DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Bacterial Samples
Specifications
Store Zirconia Beads, DNase I, 10X DNase I Buffer and DNase Inactivation Reagent at -20°C. Store RNAwiz, Wash Solution 1 and Wash Solution 2⁄3 Concentrate at 4°C. Store 0.5mL Screw Cap Tubes, Filter Cartridges, Collection Tubes, and 1.5 mL Tubes (RNase–free) at room temperature. | |
30 min. | |
Total RNA (Bacterial) | |
Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Northern Blotting, cDNA Library Construction | |
Ambion™, RiboPure™ | |
Total RNA |
Organic Extraction, Spin Column | |
Bacteria | |
Not High-throughput Compatible (Manual) | |
50 Preps | |
Up to 10^9 cells |
For Research Use Only. Not for use in diagnostic procedures.