missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RETROscript™ Reverse Transcription Kit

Product Code. 10585595
Change view
Click to view available options
40 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10585595 40 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10585595 Supplier Invitrogen™ Supplier No. AM1710

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • 50μL Control Template RNA (0.5 mg/ml)
  • 200μL 10X RT Buffer
  • 400μL 10X PCR Buffer
  • 300μL dNTP mix (2.5 mM each dNTP)
  • 80μL Random Decamers (50 μM)
  • 80μL Oligo(dT) Primers (50μL)
  • 40μL MMLV-RT (100 units/μL)
  • 40μL RNase Inhibitor (10 units/μL)
  • 50μL Control PCR Primers (5 μM each)
  • 1.0mL High Resolution Gel Loading Solution (5X)
  • 3.5mL Nuclease-free Water. 40 reactions.

Includes reagents for reverse transcription of RNA into double-stranded cDNA products

  • Amplify specific RNA targets from complex mixtures (i.e., total cellular RNA)
  • Both random decamers and oligo(dT)18 primers provided
  • Perform either one- or two-step RT-PCR
  • Optimal reaction temperature: 44° to 55°C
  • Final product size: 7kb or less

Accessory products: SuperTaq Polymerase (Mfr. Nos. AM2054 and AM2056) and SuperTaq Plus Polymerase (Mfr. Nos. AM2054 and AM2056) are recommended for use with the RETROscript Kit. RETROscript Kit components that are available separately include dNTPS for cDNA Probe Synthesis (Mfr. No. AM8200), Oligo (dT) Primer (Mfr. No. AM5730G), and Random Decamers (Mfr. No. AM5722G). Also available are the T7-Oligo (dT)24 Promoter Primer (Mfr. No. AM5710) and the T7-Oligo (dT)24 Promoter Primer, Anchored (Mfr. No. AM5712).

PCR & Real-Time PCR, Reverse Transcription

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 50μL Control Template RNA (0.5mg/mL); 200μL 10X RT Buffer; 400μL 10X PCR Buffer; 300μL dNTP mix (2.5mM each dNTP); 80μL Random Decamers (50μM); 80μL Oligo(dT) Primers (50μL); 40μL MMLV-RT (100 units/μL); 40μL RNase Inhibitor (10 units/μL); 50μL Control PCR Primers (5μM each); 1.0mL High Resolution Gel Loading Solution (5X); 3.5mL Nuclease-free Water. Store Control Template RNA below -70deg.C. Store all other reagents at -20deg.C.
Format Separate Components
Quantity 40 reactions
For Use With (Application) cDNA Synthesis, RT-PCR
Sample Type RNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.