missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ QuantumRNA™ β-Actin Internal Standards
Used for quantitative end-point RT-PCR
Brand: Invitrogen™ AM1720
Code : 50
Additional Details : Weight : 0.01000kg
Description
The Invitrogen™ QuantumRNA™ β-actin Internal Standards with Competimer™ technology uses specially modified Competimer™s of the same sequence as the normal β-actin primers that cannot be extended. By adjusting the ratio of β-actin Competimer™s to normal β-actin mRNA primers, the signal for β-actin mRNA can be attenuated to the level of very rare messages. They are used for quantitative end-point RT-PCR and include sufficient reagents for 100 reactions. β-actin mRNA is an abundantly expressed &ldquohousekeeping” gene; therefore, it is difficult to multiplex primers for β-actin mRNA in the same PCR reaction with primers for rare messages. Typically the β-actin mRNA signal will overwhelm the relative RT-PCR reaction, and the rare gene-specific product goes undetected. β-actin Standards Product Size: 294 bpSpecifications
β-Actin Primer Pair and β-Actin Competimer™s are stored at –20°C. Control Template RNA should be stored below –70°C. Nuclease-free water may be stored at any temperature. | |
Tube(s) | |
Unlabeled | |
Dry Ice | |
β-actin | |
294 bp |
Total RNA from Mouse Liver | |
ACTB | |
Ambion™, QuantumRNA™ | |
Mouse | |
100 Reactions |