missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Total RNA Blood Kit

Product Code. 10605793
Change view
Click to view available options
Quantity:
50 preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10605793 50 preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10605793 Supplier Invitrogen™ Supplier No. K156001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

PureLink Total RNA Blood Kit allows purification of high-quality total RNA from 50 to 500 μL fresh, whole mammalian blood, using non-phenol based purification in a simple spin column based protocol.

PureLink Total RNA Blood Kit allows purification of high-quality total RNA from 50 to 500 μL fresh, whole mammalian blood, using non-phenol based purification in a simple spin column based protocol.

Simple RNA Purification from Whole Blood
PureLink Total RNA Blood Kit is designed for purifying from 50 to 500 μL fresh whole blood or approximately 3.5 x 106 leukocytes (white blood cells) in less than 1 hour. The entire protocol can be done in spin tubes and spin columns.

Leukocyte Enrichment for Improved RNA Purification
Whole blood consists of non-nucleated erythrocytes (red blood cells, 4 - 6 million/mm3) and leukocytes, with the latter present in the minority (5000 - 7000/mm3). Purifying total RNA from whole blood without the removal of erythrocytes results in low RNA yields and clogging of purification columns. The distinct feature of the PureLink Total RNA Blood Kit is that it employs a leukocyte enrichment step. Hypotonic shock is used to selectively lyse erythrocytes while not affecting leukocytes. Non-nucleated erythrocytes are efficiently depleted, thus significantly reducing cell debris that may otherwise interfere with RNA purification and other downstream applications. Consequently, yields of high-quality total RNA are obtained using the PureLink Total RNA Blood Kit.

  • RNA purification from whole blood in under an hour
  • Yields high-quality RNA devoid of non-nucleated erythrocytes

Purified RNA is Suitable for Use with a Variety of Downstream Applications and Products:

  • Northern Blotting Kits & Reagents
  • Nuclease protection assays
  • Microarray
  • cDNA synthesis
  • Real-Time PCR (qPCR)

Not high-throughput compatible (Manual)

Applications
DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Blood

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 17.5 mL Lysis Buffer L3; store at room temperature
• 175 mL Lysis Buffer L5; room temperature
• 60 mL Wash Buffer W4; room temperature
• 10 mL Wash Buffer W5; room temperature
• 5 mL RNase-free Water; room temperature
• 50 Spin Cartridges with Collection Tubes; room temperature
• 50 Wash Tubes (2 mL); room temperature
• 50 Elution Tubes (1.7 mL); room temperature
Isolation Technology Silica Spin Column
Purification Time 20 min. (After sample homogenization)
Elution Volume 30–100 μL
Sample Type Blood
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 50 preps
Scale Mini
Shipping Condition Room Temperature
Starting Material Amount Blood: ≤500 μL
Yield 1000 μg (Binding capacity)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.