Learn More
Caspr2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA577698
Product Code. 15914205
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
This gene encodes a member of the neurexin family which functions in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, thrombospondin N-terminal-like domains and a putative PDZ binding site. This protein is localized at the juxtaparanodes of myelinated axons and associated with potassium channels. It may play a role in the local differentiation of the axon into distinct functional subdomains. This gene encompasses almost 1.5% of chromosome 7 and is one of the largest genes in the human genome. It may represent a positional candidate gene for the DFNB13 form of nonsyndromic deafness.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Caspr2
Polyclonal
Unconjugated
CNTNAP2
CNTNAP2, Contactin -associated protein 2
Rabbit
Antigen affinity chromatography
RUO
26047, 500105, 66797
-20°C
Lyophilized
Immunofluorescence, Immunohistochemistry, Western Blot
0.4 mg/mL
PBS with 1% BSA and 0.05% sodium azide; pH 7.4
Q9CPW0, Q9UHC6
Cntnap2
Peptide (C)DPNFTETIDESKKEWLI, corresponding to amino acid residues 1315-1331 of human Caspr2
50 μL
Primary
Human, Mouse, Rat