Learn More
Invitrogen™ PureLink™ Plant RNA Reagent
Plant RNA Reagent is a single-component solution optimized for the isolation of high yields of high-quality total RNA from a variety of plant tissues, especially those rich in polyphenolics (e.g., conifer needles) or starch (e.g., potato tuber or seeds).
Brand: Invitrogen™ 12322012
397.50 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 60-24-2 Weight : 0.10000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : II
Includes: Supplied as a ready-to-use solution.
Description
Plant RNA Reagent is a single-component solution optimized for the isolation of high yields of high-quality total RNA from a variety of plant tissues, especially those rich in polyphenolics (e.g., conifer needles) or starch (e.g., potato tuber or seeds).
Improved quality and purity of plant RNA
Plant RNA Reagent is optimized to have less sample DNA contamination, to be twice as effective as TRIzol Reagent, and have a simplified RNA extraction protocol for minimized time and labor. The end result is high-yield, high-purity RNA (determined by A260/A280 and gel analysis) from a variety of plant material sources.
Works for milligram to gram quantities of starting material
Plant RNA Reagent provides greater flexibility for sample size and processing formats, and can be used for both small-scale (up to 0.1 g plant tissue) and large-scale (0.1–5 g plant tissue) total RNA isolation. The amount supplied (100 mL) is sufficient for 200 reactions using 100 mg of tissue, or 4 reactions using 5g of tissue.
- Isolate high-quality RNA from difficult plant materials
- Use for small-scale and large-scale RNA isolations
- Fewer resources used, less hazardous, less waste
Isolated RNA is ideal for a variety of applications and products:
- cDNA library construction kits
- Real-Time PCR (qPCR)
- Northern blotting kits and reagents
- Poly(A) selection kit RNase control reagents and assays
Not high-throughput compatible (Manual)
Specifications
PureLink™ Plant RNA Reagent is supplied as a ready-to-use solution. Store at +4°C. | |
1 hr. | |
Total RNA | |
Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction, Nuclease Protection Assays, Northern Blotting | |
1 | |
Up to 1 g tissue, Up to 1 x 10^7 cells |
Organic Extraction | |
Plant Samples | |
Not High-throughput Compatible (Manual) | |
200 Preps | |
Scaleable | |
Total RNA |
Safety and Handling
- PureLink Plant RNA Reagent
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P314-Get medical advice/attention if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2-Mercaptoethanol, SODIUM AZIDE, lgepal
For Research Use Only. Not for use in diagnostic procedures.