missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ miRNA Isolation Kit

Product Code. 10605983
Change view
Click to view available options
Quantity:
25 preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10605983 25 preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10605983 Supplier Invitrogen™ Supplier No. K157001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The Invitrogen PureLink miRNA Isolation Kit provides a simple, reliable, and rapid column-based method for isolating high-quality total microRNA (miRNA) from a wide variety of sources without the need for hazardous reagents such as phenol.

The Invitrogen PureLink miRNA Isolation Kit provides a simple, reliable, and rapid column-based method for isolating high-quality total microRNA (miRNA) from a wide variety of sources without the need for hazardous reagents such as phenol. The kit provides a rapid and convenient method for purifying small RNA molecules from animal and plant cells, as well as from bacteria and yeast samples.

  • Purify small RNA molecules with a novel enrich/bind/elute procedure
  • Prepare enriched microRNA samples typically in less than 15 minutes
  • Minimize contaminating large RNA molecules and genomic DNA

Rapid and Convenient Column Purification Procedure for small RNAs
The Invitrogen PureLink miRNA Isolation Kit is specifically designed to purify small RNA molecules, including miRNAs and siRNAs, from a variety of samples typically in less than 15 minutes. The kit utilizes a silica-based two-column system to enrich exclusively for small RNAs. RNAs greater than approximately 200 nucleotides are bound to the first column. Ethanol is then added to the flow-through in order to increase the binding affinity of small RNAs to the glass fiber membrane in the second column. Once bound to the second column, small RNA molecules are then eluted, ready-for-use, and free of large RNA molecules such as 28S rRNA, 18S rRNA, and mRNA, allowing for focused expression analysis studies of small RNA molecules

Purified RNA is Suitable for Use with a Variety of Downstream Applications and Products:
The miRNA purified using the Invitrogen PureLink miRNA Isolation Kit is suitable for downstream applications including RNA sequencing, microarray analysis, real-time PCR (qPCR), reverse transcription real-time PCR (RT-qPCR), and northern blotting

Not high-throughput compatible (Manual)

Applications
DNA and RNA Purification and Analysis, RNA Extraction, RNAi, RNAi, Epigenetics and Non-Coding RNA Research, miRNA and Non-Coding RNA Analysis, miRNA Isolation, microRNA Isolation, siRNA

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 45 mL Binding Buffer L3; store at room temperature
• 10 mL Wsh Buffer W5; room temperature
• 3 mL Buffer W4; room temperature
• 5 mL RNase-free Water; room temperature
• 50 Spin Cartridges with Collection Tubes; room temperature
• 50 Wash Tubes (2 mL); room temperature
• 50 Recovery Tubes (1.7 mL); room temperature
Isolation Technology Silica Spin Column
Purification Time 20 min. (After sample homogenization)
Elution Volume 50–100 μL
Sample Type Bacteria, Cells, Plant, Tissue, Yeast, Dicing Reaction
Final Product Type Small RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) qRT-PCR, Microarray Analysis, Northern Blotting
Quantity 25 preps
Scale Mini
Shipping Condition Room Temperature
Starting Material Amount Bacteria: ≤106 cells
Cells: 106
Plant: varies
Tissue: ≤100 mg
Yeast: 1 mL
Dicing Reaction: 300 μL
Yield Up to 6 μg
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.